Tributyrin ≥97.0%
Leverantör: TCI EUROPE
Synonyms: Tributyrin, Glyceryl tributyrate, Glycerol tributyrate, Propane-1,2,3-triyl tributyrate
T0364-25ML T0364-500ML
TCIAT0364-25MLEA 245 SEK
TCIAT0364-25ML TCIAT0364-500ML
Tributyrin ≥97.0%
Tributyrin
Formel: C₁₅H₂₆O₆
MW: 302,37 g/mol
Kokpunkt: 190°C
Smältpunkt: -75°C
Densitet: 1,035 g/cm³ (20 °C)
Flampunkt: 180°C
Storage Temperature: Omgivande
MDL Number: MFCD00009392
CAS nummer: 60-01-5
EINECS: 200-451-5
Merck Index: 14,09620
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f49c36d19f7dfde3734c1fee03c59fc28b6c4b9b&vid=f49c36d19f7dfde3734c1fee03c59fc28b6c4b9b&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %