Adonitol ≥97.0%

Leverantör: TCI EUROPE

Synonyms: (2R,3s,4S)-Pentane-1,2,3,4,5-pentol, Pentane-1,2,3,4,5-pentol, (2R,3s,4S)-Pentane-1,2,3,4,5-pentaol, Adonit, Ribitol, 1,2,3,4,5-Pentanol, 1,2,3,4,5-Pentanepentol, Adonite, Pentitol, Adonitrol

A0171-1G A0171-25G
TCIAA0171-1GEA 215 SEK
TCIAA0171-1G TCIAA0171-25G
Adonitol ≥97.0%
Adonitol
Formel: HOH₂C(CH(OH))₃CH₂OH
MW: 152,15 g/mol
Smältpunkt: 100…103 °C
MDL Number: MFCD00064291
CAS nummer: 488-81-3
EINECS: 207-685-7
Merck Index: 14,00168
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0e0c6cc222f85e38c66d06ee1dbde93049d0058&vid=d0e0c6cc222f85e38c66d06ee1dbde93049d0058&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 101.0 to 104.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR