Metyl(±)-laktat ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Methyl Lactate, Methyl DL-lactate, Lactic acid methyll ester, Methyl (±)-2-hydroxypropionate

L0004-25ML L0004-500ML
TCIAL0004-25MLEA 245 SEK
TCIAL0004-25ML TCIAL0004-500ML
Metyl(±)-laktat ≥98.0%
Metyl(±)-laktat
Formel: C₄H₈O₃
MW: 104,11 g/mol
Kokpunkt: 144°C
Densitet: 1,118 g/cm³ (0 °C)
Flampunkt: 54°C
MDL Number: MFCD00066367
CAS nummer: 547-64-8
EINECS: 208-930-0
UN: 3272
ADR: 3,III
Merck Index: 14,06094
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73d0cfa7773853ae506eef3631b80cec84dd5c74&vid=73d0cfa7773853ae506eef3631b80cec84dd5c74&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR