2-Chloro-5-methylthiophene ≥96.0%
Leverantör: TCI
Varning
C2324-25G C2324-5G
TCIAC2324-25GEA 2800 SEK
TCIAC2324-25G TCIAC2324-5G
2-Chloro-5-methylthiophene ≥96.0%
2-Chloro-5-methylthiophene
Formel: C₅H₅ClS
MW: 132.61 g/mol
Kokpunkt: 155°C
MDL Number: MFCD01632145
CAS nummer: 17249-82-0
UN: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3a5d0f5117c70ca0df44550409afd309f78c158&vid=e3a5d0f5117c70ca0df44550409afd309f78c158&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %