4-Bromo-2-methylthiophene ≥98.0%

Leverantör: TCI EUROPE

B3385-25G B3385-5G
TCIAB3385-25GEA 2475 SEK
TCIAB3385-25G TCIAB3385-5G
4-Bromo-2-methylthiophene ≥98.0%
4-Bromo-2-methylthiophene
Formel: C₅H₅BrS
MW: 177,06 g/mol
Kokpunkt: 62°C
Flampunkt: 68°C
MDL Number: MFCD04116367
CAS nummer: 29421-92-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd678eb7e1efb123c30587aaa41a206d97779f9c&vid=bd678eb7e1efb123c30587aaa41a206d97779f9c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR