2-Methoxythiophene ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
M1030-25G
M1030-5G
TCIAM1030-25GEA
4825
SEK
TCIAM1030-25G
TCIAM1030-5G
2-Methoxythiophene ≥98.0%
2-Methoxythiophene
Formel:
C₅H₆OS MW: 114,17 g/mol Kokpunkt: 154°C Smältpunkt: –5 °C Densitet: 1,1335 g/cm³ (20 °C) Flampunkt: 27°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00014526 CAS nummer: 16839-97-7 EINECS: 240-863-2 UN: 1993 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...