Octamethyltrisiloxane ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Dimethylbis(trimethylsilyloxy)silane, Poly(dimethylsiloxane), 1,1,1,3,3,5,5,5-Octamethyltrisiloxane

O0257-100ML O0257-25ML
TCIAO0257-100MLEA 1275 SEK
TCIAO0257-100ML TCIAO0257-25ML
Octamethyltrisiloxane ≥98.0%
Octamethyltrisiloxane
Formel: [(CH₃)₃SiO]₂Si(CH₃)₂
MW: 236,53 g/mol
Kokpunkt: 153°C
Smältpunkt: –82 °C
Densitet: 0,82 g/cm³ (20 °C)
Flampunkt: 39°C (closed cup)
MDL Number: MFCD00008264
CAS nummer: 107-51-7
EINECS: 203-497-4
UN: 1993
ADR: 3,III
Merck Index: 14,06748
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46eaaca8ad281528ad746bb60264e269a9c1ba95&vid=46eaaca8ad281528ad746bb60264e269a9c1ba95&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR