1-Phenylpyrrole ≥98.0%

Leverantör: TCI EUROPE

Synonyms: N-Phenylpyrrole, 1-Phenyl-1H-pyrrole

P1122-10G P1122-25G P1122-5G
TCIAP1122-10GEA 1300 SEK
TCIAP1122-10G TCIAP1122-25G TCIAP1122-5G
1-Phenylpyrrole ≥98.0%
1-Phenylpyrrole
Formel: C₁₀H₉N
MW: 143,19 g/mol
Kokpunkt: 131°C
Smältpunkt: 63…64 °C
MDL Number: MFCD00005343
CAS nummer: 635-90-5
EINECS: 211-242-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09eb4b2a696acd6326c670a2fd56e31046d7a73d&vid=09eb4b2a696acd6326c670a2fd56e31046d7a73d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 58.0 to 61.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR