Pyrrol ≥99.0%
Leverantör: TCI
|
Fara
|
P0574-25ML
P0574-500ML
P0574-100ML
TCIAP0574-25MLEA
230
SEK
TCIAP0574-25ML
TCIAP0574-500ML
TCIAP0574-100ML
Pyrrol ≥99.0%
Pyrrole
Formel:
C₄H₅N Kokpunkt: 130°C Smältpunkt: -23°C Densitet: 0,97 g/cm³ (20 °C) Flampunkt: 39°C |
MDL Number:
MFCD00005216 CAS nummer: 109-97-7 EINECS: 203-724-7 UN: 1992 ADR: 3,III Merck Index: 14,08014 |
|
Specifikation testresultat
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 99.0 % |
Absorbance | max. 0.200 (400nm) |
Absorbance2 | max. 0.020 (600nm) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...