Metylnikotinat ≥98.0%
Leverantör: TCI
|
Varning
|
Synonyms:
3-Picolinic acid methyl ester, 3-(Methoxycarbonyl)pyridine, Pyridine-3-carboxylic acid methyl ester, Nicotinic acid methyl ester, 3-Pyridinecarboxylic acid methyl ester, Methyl 3-pyridinecarboxylate
N0086-25G
N0086-500G
TCIAN0086-25GEA
255
SEK
TCIAN0086-25G
TCIAN0086-500G
Metylnikotinat ≥98.0%
Metylnikotinat
Formel:
C₇H₇NO₂ Kokpunkt: 209 °C (1013 hPa) Smältpunkt: 39 °C Densitet: 1,0048 g/cm³ (20 °C) Flampunkt: 95 °C Storage Temperature: Kylskåp |
MDL Number:
MFCD00006388 CAS nummer: 93-60-7 EINECS: 202-261-8 Merck Index: 14,06100 |
|
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 39.0 to 43.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...