Isonikotinoylhydrazid ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: 4-Picolinic acid hydrazide, 4-Pyridinecarboxylic acid hydrazide, Isonicotinic hydrazide, Pyridine-4-carboxylic acid hydrazide, Isonicotinic acid hydrazide
I0138-25G I0138-500G I0138-100G
TCIAI0138-25GEA 230 SEK
TCIAI0138-25G TCIAI0138-500G TCIAI0138-100G
Isonikotinoylhydrazid ≥98.0% (by HPLC, titration analysis)
Isonikotinoylhydrazid
Formel: C₆H₇N₃O
MW: 137,14 g/mol
Smältpunkt: 169…172 °C
Densitet: 1,42 g/cm³ (20 °C)
Flampunkt: ≥ 250 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006426
CAS nummer: 54-85-3
EINECS: 200-214-6
Merck Index: 14,05186
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1946f0c3a3d86d9b196f289e965ec92a006cfcb9&vid=1946f0c3a3d86d9b196f289e965ec92a006cfcb9&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency
Melting point 170.0 to 173.0 deg-C