Klormekvatklorid ≥98.0%
Leverantör: TCI
|
Fara
|
Synonyms:
2-Kloretyltrimetylammoniumklorid
C0172-25G
C0172-500G
TCIAC0172-25GEA
495
SEK
TCIAC0172-25G
TCIAC0172-500G
Klormekvatklorid ≥98.0%
Klormekvatklorid
Formel:
ClCH₂CH₂N(CH₃)₃Cl MW: 158,07 g/mol Smältpunkt: 243 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00011869 CAS nummer: 999-81-5 EINECS: 213-666-4 UN: 2811 ADR: 6.1,III Merck Index: 14,02104 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...