Trioctylfosfinoxid ≥95.0%

Leverantör: TCI
Fara

Synonyms: TOPO, Tri-n-octylphosphine Oxide, Trioctylphosphinoxide

T0504-100G T0504-25G
TCIAT0504-100GEA 965 SEK
TCIAT0504-100G TCIAT0504-25G
Trioctylfosfinoxid ≥95.0%
Trioctylfosfinoxid
Formel: C₂₄H₅₁OP
MW: 386.64 g/mol
Kokpunkt: 238°C
MDL Number: MFCD00002083
CAS nummer: 78-50-2
EINECS: 201-121-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=089530d958c26727aace3c671c17ffe394396827&vid=089530d958c26727aace3c671c17ffe394396827&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 52.0 to 56.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR