Trifenylfosfit ≥97.0%
Leverantör: TCI
|
Varning
|
T0510-25G
T0510-500G
TCIAT0510-25GEA
215
SEK
TCIAT0510-25G
TCIAT0510-500G
Trifenylfosfit ≥97.0%
Trifenylfosfit
Formel:
C₁₈H₁₅O₃P MW: 310.29 g/mol Kokpunkt: 360°C Flampunkt: 206°C Storage Temperature: Omgivande |
MDL Number:
MFCD00003032 CAS nummer: 101-02-0 EINECS: 202-908-4 UN: 3082 ADR: 9 III |
|
Specifikation testresultat
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...