Dioctyltin oxide ≥98.0% (W)
Leverantör: TCI
Synonyms: Di-n-octyltin oxide, Dioctyloxostannane
D1373-25G D1373-500G
TCIAD1373-25GEA 230 SEK
TCIAD1373-25G TCIAD1373-500G
Dioctyltin oxide ≥98.0% (W)
Dioctyltin oxide
Formel: C₁₆H₃₄OSn
Kokpunkt: 230 °C (1013 hPa)
Densitet: 1,2 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00013839
CAS nummer: 870-08-6
EINECS: 212-791-1
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=901834bda4733d29fe8c6efb5fdc5fee64e80201&vid=901834bda4733d29fe8c6efb5fdc5fee64e80201&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (Gravimetric Method) min. 98.0 %