Dibutyltenndiklorid ≥97.0%
Leverantör: TCI EUROPE
Fara
Synonyms: DBTC
D0223-25G D0223-500G D0223-100G
TCIAD0223-25GEA 215 SEK
TCIAD0223-25G TCIAD0223-500G TCIAD0223-100G
Dibutyltenndiklorid ≥97.0%
Dibutyltenndiklorid
Formel: C₈H₁₈Cl₂Sn
MW: 303,85 g/mol
Kokpunkt: 275 °C (1013 hPa)
Smältpunkt: 37…39 °C
Densitet: 1,37 to 1.4 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00000518
CAS nummer: 683-18-1
EINECS: 211-670-0
UN: 3146
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f6ae92c377156b5edb974c6080bd5852786d20a&vid=2f6ae92c377156b5edb974c6080bd5852786d20a&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Gray to Brown powder to crystaline
Purity (Argentmetric Titration) min. 97.0 %
Melting point 40.0 to 43.0 deg-C