Flourescein
Leverantör: TCI
|
Varning
|
Synonyms:
3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid
F0095-25G
F0095-500G
F0095-100G
TCIAF0095-25GEA
310
SEK
TCIAF0095-25G
TCIAF0095-500G
TCIAF0095-100G
Flourescein
Flourescein
Formel:
C₂₀H₁₂O₅ Smältpunkt: 315°C Storage Temperature: Omgivande |
MDL Number:
MFCD00005050 CAS nummer: 2321-07-5 EINECS: 219-031-8 Merck Index: 14,04159 |
|
Specifikation testresultat
Appearance | Orange to Amber to Dark red powder to crystal |
Solubility in hot EtOH (95vol%) | almost transparency |
Drying loss | max. 1.0 % |
Ignition residue (Sulfate) | max. 2.0 % |
Sensitiveness | to pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...