Xanthene-9-carboxylic acid ≥98.0%

Leverantör: TCI
Varning

X0031-25G X0031-5G
TCIAX0031-25GEA 1525 SEK
TCIAX0031-25G TCIAX0031-5G
Xanthene-9-carboxylic acid ≥98.0%
Xanthene-9-carboxylic acid
Formel: C₁₄H₁₀O₃
MDL Number: MFCD00005059
CAS nummer: 82-07-5
EINECS: 201-394-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b0cf347ebc613c05104a56d8c73b37155e0bf3b&vid=5b0cf347ebc613c05104a56d8c73b37155e0bf3b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 210.0 to 217.0 deg-C
Solubility in Methanol within almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR