2-Bromo-9-fluorenone ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 2-Bromo-9H-fluoren-9-one

B3109-25G B3109-5G
TCIAB3109-25GEA 725 SEK
TCIAB3109-25G TCIAB3109-5G
2-Bromo-9-fluorenone ≥98.0%
2-Bromo-9-fluorenone
Formel: C₁₃H₇BrO
MW: 259,1 g/mol
MDL Number: MFCD00010789
CAS nummer: 3096-56-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5aecf6ab538c4b6899946e9a4791388d5155a11&vid=e5aecf6ab538c4b6899946e9a4791388d5155a11&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 145.0 to 152.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR