Oktametylcyklotetrasiloxan ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
Volasil 244, 2,2,4,4,6,6,8,8-Octamethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane, Silicone fluid DC 244, Dow Corning 244® fluid
O0142-25ML
O0142-500ML
O0142-100ML
TCIAO0142-25MLEA
215
SEK
TCIAO0142-25ML
TCIAO0142-500ML
TCIAO0142-100ML
Oktametylcyklotetrasiloxan ≥98.0%
Oktametylcyklotetrasiloxan
Formel:
[-Si(CH₃)₂O-]₄ MW: 296,62 g/mol Kokpunkt: 175°C Smältpunkt: 18 °C Densitet: 0,95 g/cm³ (20 °C) Flampunkt: 54°C |
MDL Number:
MFCD00003269 CAS nummer: 556-67-2 EINECS: 209-136-7 UN: 1992 ADR: 3,III Merck Index: 14,06747 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...