Etynylcyklopropan ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Cyclopropylacetylene

C1984-25G C1984-5G
TCIAC1984-25GEA 1525 SEK
TCIAC1984-25G TCIAC1984-5G
Etynylcyklopropan ≥98.0%
Etynylcyklopropan
Formel: C₅H₆
MW: 66,1 g/mol
Kokpunkt: 52°C
Flampunkt: -17°C
MDL Number: MFCD02181090
CAS nummer: 6746-94-7
EINECS: 425-430-1
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=549597246b1b4f65be87659a9bbd072233ab373b&vid=549597246b1b4f65be87659a9bbd072233ab373b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR