Bifenyl-2,2'-diol ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 2,2'-Biphenol, 2,2-Biphenol, Biphenyl-2,2'-diol

B0813-25G B0813-500G B0813-100G
TCIAB0813-25GEA 510 SEK
TCIAB0813-25G TCIAB0813-500G TCIAB0813-100G
Bifenyl-2,2'-diol ≥98.0%
Bifenyl-2,2'-diol
Formel: C₁₂H₁₀O₂
MW: 186,21 g/mol
Kokpunkt: 325°C
Smältpunkt: 106…109 °C
Densitet: 1,342 g/cm³ (20 °C)
MDL Number: MFCD00002210
CAS nummer: 1806-29-7
EINECS: 217-303-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7892850df32fc743825b01a57b463affe4f9582&vid=b7892850df32fc743825b01a57b463affe4f9582&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 107.0 to 111.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR