4,4'-Di-tert-butylbiphenyl ≥98.0%
Leverantör: TCI EUROPE
Synonyms: 1-tert-butyl-4-(4-tert-butylphenyl)benzene, 4,4'-Di-tert-butyl-1,1'-biphenyl
D2450-25G D2450-5G
TCIAD2450-25GEA 1925 SEK
TCIAD2450-25G TCIAD2450-5G
4,4'-Di-tert-butylbiphenyl ≥98.0%
4,4'-Di-tert-butylbiphenyl
Formel: C₂₀H₂₆
MW: 266,43 g/mol
Kokpunkt: 228 °C (1013 hPa)
Smältpunkt: 127 °C
Densitet: 0,9691 g/cm³ (20 °C)
MDL Number: MFCD00008834
CAS nummer: 1625-91-8
EINECS: 216-615-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5edeb06531bdf830daa6aa3f12c1b18e8ab89eb&vid=e5edeb06531bdf830daa6aa3f12c1b18e8ab89eb&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 126.0 to 130.0 deg-C
Solubility in Toluene almost transparency