4-Ethylbenzyl alcohol ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: (4-Ethylphenyl)methanol

E0704-25G E0704-5G
TCIAE0704-25GEA 3025 SEK
TCIAE0704-25G TCIAE0704-5G
4-Ethylbenzyl alcohol ≥98.0%
4-Ethylbenzyl alcohol
Formel: C₉H₁₂O
MW: 136,19 g/mol
Kokpunkt: 117°C
MDL Number: MFCD00004666
CAS nummer: 768-59-2
EINECS: 212-198-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a24cde86f03d482303d7d583342684b0c8ccc73c&vid=a24cde86f03d482303d7d583342684b0c8ccc73c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR