4-Pentylbiphenyl ≥96.0%

Leverantör: TCI EUROPE

Synonyms: 4-Amylbiphenyl, 4-n-Pentylbiphenyl

P1133-25ML P1133-5ML
TCIAP1133-25MLEA 1800 SEK
TCIAP1133-25ML TCIAP1133-5ML
4-Pentylbiphenyl ≥96.0%
4-Pentylbiphenyl
Formel: C₁₇H₂₀
MW: 224,35 g/mol
Kokpunkt: 167°C
Smältpunkt: 9,5…10.5 °C
Densitet: ~ 0,94 g/cm³ (20 °C)
Flampunkt: ≥ 110 °C
MDL Number: MFCD00009503
CAS nummer: 7116-96-3
EINECS: 230-421-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2dd510e0b738e7044f73ca5e115eab9df720f889&vid=2dd510e0b738e7044f73ca5e115eab9df720f889&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Orange to Green clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR