2-Methyl-1-phenyl-2-propanol ≥98.0%

Leverantör: TCI EUROPE
Varning

D0783-25ML D0783-500ML D0783-100ML
TCIAD0783-25MLEA 215 SEK
TCIAD0783-25ML TCIAD0783-500ML TCIAD0783-100ML
2-Methyl-1-phenyl-2-propanol ≥98.0%
2-Methyl-1-phenyl-2-propanol
Formel: C₆H₅CH₂C(CH₃)(OH)CH₃
MW: 150,22 g/mol
Kokpunkt: 94°C
Smältpunkt: 23…25 °C
Densitet: 0,974 g/cm³ (20 °C)
MDL Number: MFCD00004465
CAS nummer: 100-86-7
EINECS: 202-896-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac777db219c13ec81e86506d935e238c9a78e573&vid=ac777db219c13ec81e86506d935e238c9a78e573&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR