2-Ethoxybenzyl alcohol ≥98.0%
Leverantör: TCI EUROPE
Synonyms: (2-Ethoxyphenyl)methanol
E0499-25G E0499-5G
TCIAE0499-25GEA 5425 SEK
TCIAE0499-25G TCIAE0499-5G
2-Ethoxybenzyl alcohol ≥98.0%
2-Ethoxybenzyl alcohol
Formel: C₉H₁₂O₂
MW: 152,19 g/mol
Kokpunkt: 265°C
Smältpunkt: 0 °C
Densitet: 1,07 g/cm³ (20 °C)
MDL Number: MFCD00004616
CAS nummer: 71672-75-8
EINECS: 275-811-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00d78456e09e24f935314c971ef8a9eedfb7c770&vid=00d78456e09e24f935314c971ef8a9eedfb7c770&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %