2-Aminobenzyl alcohol ≥98.0% (by GC, titration analysis)
Leverantör: TCI
Varning
Synonyms: (2-Aminophenyl)methanol, 2-Hydroxymethylaniline, 2-(Hydroxymethyl)aniline, 2-Aminobenzyl Alcohol
A1000-25G A1000-250G
TCIAA1000-25GEA 735 SEK
TCIAA1000-25G TCIAA1000-250G
2-Aminobenzyl alcohol ≥98.0% (by GC, titration analysis)
2-Aminobenzyl alcohol
Formel: C₇H₉NO
MW: 123.15 g/mol
Kokpunkt: 275
Smältpunkt: 81
Storage Temperature: Kylskåp
MDL Number: MFCD00007749
CAS nummer: 5344-90-1
EINECS: 226-293-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c557b80b77f59a6d23a0c27dbe91897a2ef89ff&vid=0c557b80b77f59a6d23a0c27dbe91897a2ef89ff&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Orange to Green powder to crystalline
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 82.0 to 86.0 deg-C