Fenetylamin ≥98.0% (by GC, titration analysis)
Leverantör: TCI
Fara
Synonyms: PEA, β-Phenylethylamine, 2-Phenylethylamine
P0085-25ML P0085-500ML P0085-100ML
TCIAP0085-25MLEA 215 SEK
TCIAP0085-25ML TCIAP0085-500ML TCIAP0085-100ML
Fenetylamin ≥98.0% (by GC, titration analysis)
Fenetylamin
Formel: C₆H₅CH₂CH₂NH₂
MW: 121.18 g/mol
Kokpunkt: 200
Smältpunkt: -60
Flampunkt: 90°C
Storage Temperature: Omgivande
MDL Number: MFCD00008184
CAS nummer: 64-04-0
EINECS: 200-574-4
UN: 2735
ADR: 8 II
Merck Index: 14,06026
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c0fd4fcc97e2e8069a5894ff4ad5e16ccae117d&vid=9c0fd4fcc97e2e8069a5894ff4ad5e16ccae117d&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %