Benzyl(dimetyl)amin ≥98.0%
Leverantör: TCI
|
Fara
|
D0688-25ML
D0688-500ML
TCIAD0688-25MLEA
215
SEK
TCIAD0688-25ML
TCIAD0688-500ML
Benzyl(dimetyl)amin ≥98.0%
Benzyl(dimetyl)amin
Formel:
C₆H₅CH₂N(CH₃)₂ Kokpunkt: 180°C Smältpunkt: –75°C Densitet: 0,915 g/cm³ (20 °C) Flampunkt: 57°C Storage Temperature: Omgivande |
MDL Number:
MFCD00008329 CAS nummer: 103-83-3 EINECS: 203-149-1 UN: 2619 ADR: 8,II |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...