4-(Trifluoromethyl)benzenesulphonyl chloride ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: 4-(Trifluoromethyl)benzene-1-sulphonyl chloride

T1795-1G T1795-5G
TCIAT1795-1GEA 230 SEK
TCIAT1795-1G TCIAT1795-5G
4-(Trifluoromethyl)benzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
4-(Trifluoromethyl)benzenesulphonyl chloride
Formel: C₇H₄ClF₃O₂S
MW: 244,62 g/mol
Kokpunkt: 58°C
Smältpunkt: 31…33 °C
MDL Number: MFCD00042422
CAS nummer: 2991-42-6
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b8b41fe5e3a2cc9ad0e794c7573401fe2019e65&vid=2b8b41fe5e3a2cc9ad0e794c7573401fe2019e65&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorles to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR