2-(Trifluoromethoxy)benzyl alcohol ≥96.0%
Leverantör: TCI
|
Varning
|
T2593-1G
T2593-5G
TCIAT2593-1GEA
550
SEK
TCIAT2593-1G
TCIAT2593-5G
2-(Trifluoromethoxy)benzyl alcohol ≥96.0%
2-(Trifluoromethoxy)benzyl alcohol
Formel:
C₈H₇F₃O₂ MW: 192.14 g/mol |
MDL Number:
MFCD00153285 CAS nummer: 175278-07-6 |
|
Specifikation testresultat
Appearance | White to Almost white clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...