4-Fluoro-3-nitrotoluene ≥98.0%
Leverantör: TCI
Varning
F0562-25G F0562-5G
TCIAF0562-25GEA 1425 SEK
TCIAF0562-25G TCIAF0562-5G
4-Fluoro-3-nitrotoluene ≥98.0%
4-Fluoro-3-nitrotoluene
Formel: C₇H₆FNO₂
MW: 155.13 g/mol
Kokpunkt: 241°C
MDL Number: MFCD00007060
CAS nummer: 446-11-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e38390b1213d1d112fe89a9b19271e464c97e943&vid=e38390b1213d1d112fe89a9b19271e464c97e943&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light orange to Yellow to Green powder to lump
Purity (GC) min. 98.0 %