2',4'-Difluoropropiophenone ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 1-(2,4-Difluorophenyl)propan-1-one

D2598-25G D2598-5G
TCIAD2598-25GEA 1900 SEK
TCIAD2598-25G TCIAD2598-5G
2',4'-Difluoropropiophenone ≥98.0%
2',4'-Difluoropropiophenone
Formel: C₉H₈F₂O
MW: 170,16 g/mol
Kokpunkt: 52 °C (1013 hPa)
Densitet: 1,193 g/cm³ (20 °C)
Flampunkt: 76°C
MDL Number: MFCD00015507
CAS nummer: 85068-30-0
EINECS: 285-291-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4419cc9a359c1530c9dea8f2dc98b1a47cd946d1&vid=4419cc9a359c1530c9dea8f2dc98b1a47cd946d1&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR