4'-Hydroxypropiophenone ≥98.0%
Leverantör: TCI
|
Varning
|
Synonyms:
4’-Hydroxypropiophenone, Paroxypropione, p-Hydroxypropiophenone
H0299-25G
H0299-500G
TCIAH0299-25GEA
215
SEK
TCIAH0299-25G
TCIAH0299-500G
4'-Hydroxypropiophenone ≥98.0%
4'-Hydroxypropiophenone
Formel:
C₉H₁₀O₂ MW: 150,18 g/mol Kokpunkt: 140 - 145 °C (0,5 -rr) Smältpunkt: 147…150 °C Densitet: 1,09 g/cm³ (20 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00002361 CAS nummer: 70-70-2 EINECS: 200-743-2 Merck Index: 14,07044 |
|
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 148.0 to 152.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...