3'-Chloropropiophenone ≥98.0%

Leverantör: TCI EUROPE

Synonyms: m-Chloropropiophenone, 1-(3-Chlorophenyl)propan-1-one

C2033-250G C2033-25G
TCIAC2033-250GEA 2450 SEK
TCIAC2033-250G TCIAC2033-25G
3'-Chloropropiophenone ≥98.0%
3'-Chloropropiophenone
Formel: C₉H₉ClO
MW: 168,62 g/mol
Kokpunkt: 124°C
Smältpunkt: 43…47 °C
MDL Number: MFCD00009925
CAS nummer: 34841-35-5
EINECS: 252-242-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46a5090045db0c50a683865980f050238a3ee949&vid=46a5090045db0c50a683865980f050238a3ee949&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 45.0 to 49.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR