5-Bromo-2-iodotoluene ≥98.0%

Leverantör: TCI
Varning

Synonyms: 1-Bromo-4-iodo-3-methylbenzene

B2069-25G B2069-5G
TCIAB2069-25GEA 1350 SEK
TCIAB2069-25G TCIAB2069-5G
5-Bromo-2-iodotoluene ≥98.0%
5-Bromo-2-iodotoluene
Formel: C₇H₆BrI
MW: 296.93 g/mol
Kokpunkt: 260°C
MDL Number: MFCD00060664
CAS nummer: 116632-39-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6165b3c51ffa33eae8a77f7b5380dd54b89f492b&vid=6165b3c51ffa33eae8a77f7b5380dd54b89f492b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR