ω,4-Dibromoacetophenone ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Tau Protein Kinase I Inhibitor, 2,4'-Dibromoacetophenone, Glycogen Synthase Kinase-3β Inhibitor VII, α,4-Dibromoacetophenone, GSK-3β Inhibitor VII, TPK I Inhibitor, 2-Bromo-1-(4-bromophenyl)-1-ethanone, 4-Bromophenacyl bromide

D0164-25G D0164-100G
TCIAD0164-25GEA 525 SEK
TCIAD0164-25G TCIAD0164-100G
ω,4-Dibromoacetophenone ≥98.0%
ω,4-Dibromoacetophenone
Formel: C₈H₆Br₂O
MW: 277,94 g/mol
Kokpunkt: 310 °C (1013 hPa)
Smältpunkt: 107…108 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00000200
CAS nummer: 99-73-0
EINECS: 202-783-6
UN: 3261
ADR: 8,III
Merck Index: 14,01427
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=572ce097b6aa1d8ad12e47dd8f6c3c407e652875&vid=572ce097b6aa1d8ad12e47dd8f6c3c407e652875&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 109.0 to 113.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR