3'-Bromopropiophenone ≥98.0%

Leverantör: TCI

Synonyms: m-Bromopropiophenone, 1-(3-Bromophenyl)propan-1-one

B1888-25G B1888-5G
TCIAB1888-25GEA 4400 SEK
TCIAB1888-25G TCIAB1888-5G
3'-Bromopropiophenone ≥98.0%
3'-Bromopropiophenone
Formel: C₉H₉BrO
Kokpunkt: 145°C
Smältpunkt: 39…41 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00000084
CAS nummer: 19829-31-3
EINECS: 243-353-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a83a78862c38cd80c702584e97fa51718191f56&vid=9a83a78862c38cd80c702584e97fa51718191f56&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 39.0 to 42.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR