1-Bromo-4-propylbenzene ≥97.0%
Leverantör: TCI
|
Varning
|
Synonyms:
(4-Bromophenyl)propane, 1-Bromo-4-n-propylbenzene
B1889-25G
B1889-5G
TCIAB1889-25GEA
2600
SEK
TCIAB1889-25G
TCIAB1889-5G
1-Bromo-4-propylbenzene ≥97.0%
1-Bromo-4-propylbenzene
Formel:
C₉H₁₁Br MW: 199.09 g/mol Kokpunkt: 225°C Smältpunkt: -41.5 Flampunkt: 95°C |
MDL Number:
MFCD00012456 CAS nummer: 588-93-2 EINECS: 454-790-0 |
|
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...