1-Bromo-4-pentylbenzene ≥90.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 4-Pentylbromobenzene, 4-Bromopentylbenzene, (4-Bromophenyl)pentane

B1605-25G B1605-5G
TCIAB1605-25GEA 695 SEK
TCIAB1605-25G TCIAB1605-5G
1-Bromo-4-pentylbenzene ≥90.0%
1-Bromo-4-pentylbenzene
Formel: C₁₁H₁₅Br
MW: 227,14 g/mol
Kokpunkt: 148°C
Flampunkt: 89°C
MDL Number: MFCD00061113
CAS nummer: 51554-95-1
EINECS: 472-220-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1292c6c125d8efa1c144a3823f1ecab983e05c5&vid=d1292c6c125d8efa1c144a3823f1ecab983e05c5&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR