1-Bromo-4-nonylbenzene ≥95.0%
Leverantör: TCI
|
Varning
|
B1596-25G
B1596-5G
B1596-1G
TCIAB1596-25GEA
7750
SEK
TCIAB1596-25G
TCIAB1596-5G
TCIAB1596-1G
1-Bromo-4-nonylbenzene ≥95.0%
1-Bromo-4-nonylbenzene
Formel:
C₁₅H₂₃Br Kokpunkt: 190°C |
MDL Number:
MFCD00061116 CAS nummer: 51554-94-0 |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...