1-Bromo-4-hexylbenzene ≥90.0%

Leverantör: TCI EUROPE

Synonyms: (4-Bromophenyl)hexane

B1606-25G B1606-5G
TCIAB1606-25GEA 2125 SEK
TCIAB1606-25G TCIAB1606-5G
1-Bromo-4-hexylbenzene ≥90.0%
1-Bromo-4-hexylbenzene
Formel: C₁₂H₁₇Br
MW: 241,17 g/mol
Kokpunkt: 145°C
MDL Number: MFCD00061114
CAS nummer: 23703-22-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b2fd9cfe9bd7a8ec204bc831e5c0a673e4cd3b3&vid=5b2fd9cfe9bd7a8ec204bc831e5c0a673e4cd3b3&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR