1-Bromo-4-butylbenzene ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
(4-Bromophenyl)butane
B1907-25G
B1907-5G
TCIAB1907-25GEA
605
SEK
TCIAB1907-25G
TCIAB1907-5G
1-Bromo-4-butylbenzene ≥97.0%
1-Bromo-4-butylbenzene
Formel:
C₁₀H₁₃Br MW: 213,12 g/mol Kokpunkt: 246°C Smältpunkt: –26 °C Densitet: 1,2347 g/cm³ (20 °C) Flampunkt: 108°C |
MDL Number:
MFCD00040934 CAS nummer: 41492-05-1 EINECS: 472-210-4 |
Specifikation testresultat
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...