1-Fluoro-4-ethynylbenzene ≥98.0%

Leverantör: TCI
Varning

Synonyms: 1-Ethynyl-4-fluorobenzene, (4-Fluorophenyl)acetylene, (4-Fluorophenyl)ethyne

F0470-1G F0470-5G
TCIAF0470-1GEA 470 SEK
TCIAF0470-1G TCIAF0470-5G
1-Fluoro-4-ethynylbenzene ≥98.0%
1-Fluoro-4-ethynylbenzene
Formel: C₈H₅F
Flampunkt: 29°C
Storage Temperature: Kylskåp
MDL Number: MFCD00168823
CAS nummer: 766-98-3
UN: 1325
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5bc1019f5351696ec6cc889ccb9d09c095da7812&vid=5bc1019f5351696ec6cc889ccb9d09c095da7812&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR