Fenetol ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Ethoxybenzene, Ethyl phenyl ether, Phenoxyethane

E0043-25ML E0043-500ML
TCIAE0043-25MLEA 215 SEK
TCIAE0043-25ML TCIAE0043-500ML
Fenetol ≥98.0%
Fenetol
Formel: C₆H₅OC₂H₅
MW: 122,17 g/mol
Kokpunkt: 173°C
Smältpunkt: –30 °C
Densitet: 0,956 g/cm³ (20 °C)
Flampunkt: 57°C
Storage Temperature: Omgivande
MDL Number: MFCD00009090
CAS nummer: 103-73-1
EINECS: 203-139-7
UN: 3271
ADR: 3,III
Merck Index: 14,07229
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f68d46a525d051512916ee99cca86c0fd407817&vid=4f68d46a525d051512916ee99cca86c0fd407817&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR