2-Methoxyphenyl isothiocyanate ≥98.0%

Leverantör: TCI
Fara

I0512-25G I0512-5G
TCIAI0512-25GEA 2375 SEK
TCIAI0512-25G TCIAI0512-5G
2-Methoxyphenyl isothiocyanate ≥98.0%
2-Methoxyphenyl isothiocyanate
Formel: C₈H₇NOS
Kokpunkt: 267°C
MDL Number: MFCD00011675
CAS nummer: 3288-04-8
UN: 1760
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f41b94bd5137b146c3a69ad402f3c68b102db80&vid=8f41b94bd5137b146c3a69ad402f3c68b102db80&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR