Ethyl-4-iodobenzoate ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Ethyl 4-iodobenzoate, 4-Iodobenzoic acid ethyl ester
I0369-25G
I0369-5G
TCIAI0369-25GEA
775
SEK
TCIAI0369-25G
TCIAI0369-5G
Ethyl-4-iodobenzoate ≥98.0%
Ethyl-4-iodobenzoate
Formel:
C₉H₉IO₂ MW: 276,07 g/mol Kokpunkt: 283°C Densitet: 1,655 g/cm³ (20 °C) |
MDL Number:
MFCD00017344 CAS nummer: 51934-41-9 EINECS: 429-550-3 |
Specifikation testresultat
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...