3,4-Difluorobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: 3,4-Difluorobenzene-1-sulphonyl chloride

D2941-1G D2941-5G
TCIAD2941-1GEA 270 SEK
TCIAD2941-1G TCIAD2941-5G
3,4-Difluorobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
3,4-Difluorobenzenesulphonyl chloride
Formel: C₆H₃ClF₂O₂S
MW: 212,6 g/mol
Kokpunkt: 105 °C (15 -rr)
Densitet: 1,586 g/cm³ (20 °C)
MDL Number: MFCD00153120
CAS nummer: 145758-05-0
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00d16e46bcb4a8b728f95f01ecf191ce2ed86d50&vid=00d16e46bcb4a8b728f95f01ecf191ce2ed86d50&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR