Butyl salicylate ≥99.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Butyl 2-hydroxybenzoate, Butyl 2-Hydroxybenzoate, Salicylic acid butyl ester, 2-Hydroxybenzoic acid butyl ester

S0010-25ML S0010-500ML
TCIAS0010-25MLEA 215 SEK
TCIAS0010-25ML TCIAS0010-500ML
Butyl salicylate ≥99.0%
Butyl salicylate
Formel: C₁₁H₁₄O₃
MW: 194,23 g/mol
Kokpunkt: 260°C
Densitet: 1,08 g/cm³ (25 °C)
Flampunkt: 134°C
MDL Number: MFCD00020038
CAS nummer: 2052-14-4
EINECS: 218-142-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0a0c5159a7daf9c93044173a8e8d9c7aa6c427a&vid=c0a0c5159a7daf9c93044173a8e8d9c7aa6c427a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR